Learn More
Selleck Chemical LLC Clofarabine 50mg 123318-82-1
SDP
Supplier:  Selleck Chemical LLC S121850MG
Encompass_Preferred
Clofarabine inhibits the enzymatic activities of ribonucleotide reductase (RNR) (IC50 = 65 nM) and DNA polymerase. Clofarabine induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-674-0