Learn More
Medchemexpress LLC Clofencet 1g | 129025-54-3 | 278.69 g/mol | C13H11ClN2O3 | 1 G

Supplier: Medchemexpress LLC HY1148571G
Clofencet is a small-molecule research chemical used as a wheat hybridizing agent. It is supplied as a high-purity solid for laboratory and agricultural research and is accompanied by safety documentation for handling and storage.
- High purity: 98.62% (reported).
- Molecular weight: 278.69 g/mol.
- Chemical formula: C13H11ClN2O3.
- Available in small packaging sizes suitable for lab use, including 1 G.
- English safety data sheet provided for safe handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.