Learn More
Medchemexpress LLC HY-B0287 500mg Medchemexpress, Clofibrate CAS:637-07-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0287500MG
Encompass_Preferred
Medchemexpress, HY-B0287 500mg Clofibrate CAS:637-07-0 Clofibrate is an agonist of PPAR, with EC50s of 50 μM, ∼500 μM for murine PPARα and PPARγ, and 55 μM, ∼500 μM for human PPARα and PPARγ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2447