Learn More
Medchemexpress LLC Clofibric acid | 882-09-7 | 99.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1415
Encompass_Preferred
Clofibric acid, also known as Chlorofibrinic acid, is an orally active PPARα agonist that inhibits the fimbriation of Escherichia coli and increases superoxide dismutase (SOD) activity. This compound has been shown to lower blood lipids, prevent experimental pyelonephritis, and demonstrate anticancer activity against ovarian cancer. It is also used as a herbicide. Research applications include ovarian cancer, liver cancer, obesity, and urinary tract infections.
  • Orally active
  • Acts as a PPARα agonist
  • Inhibits Escherichia coli fimbriation
  • Increases SOD activity
  • Lowers blood lipids
  • Prevents experimental pyelonephritis
  • Demonstrates anticancer activity, specifically in ovarian cancer
  • Functions as a herbicide
  • Utilized in research for various conditions such as ovarian cancer, liver cancer, obesity, and urinary tract infections
Catalog No. 50-004-14319