Learn More
Medchemexpress LLC Clofibric acid | 882-09-7 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB14155G
Encompass_Preferred
Clofibric acid (Chlorofibrinic acid) is an orally active PPARα agonist. It inhibits the fimbriation of Escherichia coli, increases SOD activity, lowers blood lipids, and prevents experimental pyelonephritis. It also has anticancer activity against ovarian cancer and is used as a herbicide. It is used in ovarian cancer, liver cancer, obesity, and urinary tract infection research.
  • Orally active PPARα agonist.
  • Inhibits the fimbriation of Escherichia coli.
  • Increases SOD activity.
  • Lowers blood lipids and prevents experimental pyelonephritis.
  • Has anticancer activity against ovarian cancer.
  • Also a herbicide.
  • Used in ovarian cancer, liver cancer, obesity, and urinary tract infection research.
Catalog No. 50-004-12580