Learn More
Medchemexpress LLC Clofibric acid | 882-09-7 | >99.92% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1415500MG
Encompass_Preferred
Clofibric acid (Chlorofibrinic acid) is an orally active PPARα agonist that lowers blood lipids and prevents experimental pyelonephritis. It also exhibits anticancer activity against ovarian cancer and is used in research areas such as ovarian cancer, liver cancer, obesity, and urinary tract infection. Additionally, Clofibric acid functions as a herbicide.
  • Acts as an orally active PPARα agonist
  • Inhibits fimbriation of Escherichia coli
  • Increases SOD activity
  • Lowers blood lipids
  • Prevents experimental pyelonephritis
  • Exhibits anticancer activity against ovarian cancer
  • Used in research for ovarian cancer, liver cancer, obesity, and urinary tract infections
  • Functions as a herbicide
Catalog No. 50-004-13674