Learn More
Medchemexpress LLC Clofibric acid (Chlorofibrinic acid) | 882-09-7 | MFCD00004192 | 99.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB141510G
Encompass_Preferred
Clofibric acid (Chlorofibrinic acid) is an orally active PPARα agonist. It inhibits the fimbriation of *Escherichia coli*, increases SOD activity, lowers blood lipids, and prevents experimental pyelonephritis. Clofibric acid also exhibits anticancer activity against ovarian cancer and is used in ovarian cancer, liver cancer, obesity, and urinary tract infection research. It is also a herbicide.
  • Orally active PPARα agonist.
  • Inhibits *Escherichia coli* fimbriation.
  • Increases SOD activity.
  • Lowers blood lipids.
  • Prevents experimental pyelonephritis.
  • Anticancer activity against ovarian cancer.
  • Used in research for ovarian cancer, liver cancer, obesity, and urinary tract infection.
Catalog No. 50-004-12683