Learn More
Enzo Life Sciences Clofilium . tosylate (100mg). CAS: 92953-10-1
SDP
Supplier:  Enzo Life Sciences ALX550242M100
Encompass_Preferred
K+ channel blocker. Antiarrhytmic. Cardiac depressant. Alternative name: 4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium . tosylate. Purity: ≥97% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: Ambient.
Catalog No. 50-200-9507