Learn More
Medchemexpress LLC Clofoctol | 37693-01-9 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB115010MM/1ML
Encompass_Preferred
Clofoctol is a bacteriostatic antibiotic primarily used to treat respiratory tract and ear, nose, and throat infections caused by Gram-positive bacteria. It is effective against Gram-positive bacteria and can penetrate human lung tissue. Clofoctol also functions as an inhibitor of prostate cancer and demonstrates antiviral potency.
  • Inhibits prostate cancer cell growth.
  • Arrests cell cycle at G1 phase.
  • Induces ER stress and activates UPR pathways.
  • Inhibits translation.
  • Shows antiviral effect against SARS-CoV-2.
  • Capable of blocking tumor growth in mouse models.
  • Reduces viral load and inflammation in lungs during SARS-CoV-2 infection.
Catalog No. 50-004-12197