Learn More
Medchemexpress LLC Clomifene-d5 hydrochloride | 1346606-66-3 | 98.5% | C26H24D5Cl2NO | 1 MG
SDP
Supplier:  Medchemexpress LLC HYA0287S1MG
Encompass_Preferred
Clomifene-d5 (hydrochloride) is a deuterium labeled Clomifene. Clomifene is an orally active compound used for infertility. It is a simple, relatively safe, easily administered, and well-tolerated efficacious compound to induce ovulation. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound.
  • Orally active.
  • Used for infertility.
  • Simple to use.
  • Relatively safe.
  • Easily administered.
  • Well-tolerated.
  • Efficacious in inducing ovulation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).
Catalog No. 50-004-32908