Learn More
Medchemexpress LLC Clomiphene (citrate) | 50-41-9 | 98.2% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04635G
Encompass_Preferred
Clomiphene citrate is an orally active estrogen-receptor modulator. It exhibits anti-cancer activity, induces perturbations during meiotic maturation and cytogenetic abnormalities, and helps in managing psychiatric and cognitive impairment.
  • Exhibits anti-cancer activity
  • Induces perturbations during meiotic maturation and cytogenetic abnormalities
  • Ameliorates psychiatric and cognitive impairment
  • Inhibits viability of human prostate cancer cells
  • Induces chromosome abnormalities in mouse oocytes
  • Shows anxiolytic potential and improved cognitive functions in PCOS models
Catalog No. 50-004-08426