Learn More
Apexbio Technology LLC Clomipramine 303-49-1 25mg
SDP
Supplier:  Apexbio Technology LLC BA854925
Encompass_Preferred
Clomipramine (CAS 303-49-1) is a tricyclic antidepressant that functions primarily as a potent inhibitor of serotonin (5-HT) reuptake with an IC50 value of 1 5 nM By blocking the reabsorption of serotonin at the synaptic cleft clomipramine increases synaptic serotonin levels which is associated with its antidepressant and anti-obsessive effects This compound is widely utilized in preclinical and clinical research to investigate the pathophysiology and potential treatments of depressive disorders and obsessive-compulsive disorder (OCD)
Catalog No. 50-000-89523