Learn More
STEMCELL Technologies ClonaCell™-CHO CD Liquid Medium, Size: 500 mL
SDP
Supplier:  STEMCELL Technologies 03817
Encompass
ClonaCell™-CHO CD Liquid Medium is a general purpose liquid medium for culture of suspension-adapted CHO cells. This chemically defined liquid medium is protein-free and animal component-free. It does not contain L-glutamine, selection agents, antibiotics, or phenol red. ClonaCell™-CHO CD Liquid Medium is compatible with a variety of selection systems, including dihydrofolate reductase (DHFR) and glutamine synthetase (GS).
Catalog No. NC0997413
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov