Learn More
Medchemexpress LLC Clonidine | 4205-90-7 | 99.9% | 100 MG

Supplier: Medchemexpress LLC HY12721100MG
Clonidine is an alpha 2-adrenergic agonist known for its ability to suppress the firing activity of neurons. This high-purity chemical, appearing as a white to off-white solid, is suitable for various research applications.
- Alpha 2-adrenergic agonist
- Suppresses neuronal firing activity
- Purity of 99.9%
- Solid, white to off-white appearance
- Molecular formula C9H9Cl2N3
- Molecular weight 230.10
- Store at 4°C, protect from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.