Learn More
Medchemexpress LLC Cloperastine fendizoate | 85187-37-7 | 99.5% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB217910G
Encompass_Preferred
Cloperastine fendizoate inhibits the hERG K+ currents in a concentration-dependent manner with an IC50 value of 27 nM. This compound possesses antitussive and antiedemic activity, relaxing bronchial musculature. It is a drug with a central antitussive effect, endowed with an antihistaminic and papaverine-like activity, similar to codeine but without its narcotic effects.
  • Inhibits hERG K+ currents.
  • Possesses antitussive and antiedemic activity.
  • Relaxes bronchial musculature.
  • Exhibits a central antitussive effect.
  • Endowed with antihistaminic and papaverine-like activity.
  • Similar to codeine but without its narcotic effects.
Catalog No. 50-004-11621