Learn More
Formumax Scientific Inc CLOPHOSOME-A-CTRL LIPSO 10ML
Supplier:  Formumax Scientific Inc F70101CAC10
Encompass_Preferred
Combo: Clophosome®-A and Control Liposomes Included in this convenience combo kit areOne 10.0 mL of Clophosome®-A - anionic clodronate liposomes (F70101C-A-10) One 10.0 mL of control anionic liposomes (F70101-A-10)
Catalog No. NC1488571