Learn More
AdipoGen Clopidogrel bisulfate
SDP
Supplier:  AdipoGen CDXD0505M250
Encompass_Preferred
Chemical. CAS 120202-66-6. Formula C16H16ClNO2S . H2SO4. MW 321.8 . 98.1. Synthetic. S--Clopidogrel is a prodrug that is metabolized by cytochrome P450 isoforms to produce an active metabolite, which is a selective, irreversible high-affinity antagonist of the platelet purinergic P2Y12 receptor. Potent antithrombotic antiplatelet agent. By inhibiting the Purinoceptor P2Y12 subtype it consequently inhibits ADP-induced platelet aggregation. S--Clopidogrel hydrogen sulfate is the active isomer. Compound can be used as analytical reference material.
Catalog No. 50-256-1064