Learn More
Medchemexpress LLC Clopidogrel impurity 1 | 120202-71-3 | 419.89 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW010935500MG
Encompass_Preferred
Clopidogrel impurity 1 is an impurity of Clopidogrel (HY-15283) intended for research use only. This product is not sold to patients. It appears as a white to off-white solid with a molecular weight of 419.89 and a CAS number of 120202-71-3. Shipping occurs at room temperature within the continental US, and storage is recommended at 4°C under nitrogen. If stored in solvent, it can be kept at -80°C for 6 months or -20°C for 1 month, also under nitrogen.
  • Impurity of clopidogrel.
  • Suitable for research purposes only.
  • Available in various quantities from 50 mg to 1 g, with bulk inquiry options.
  • Solid appearance, white to off-white in color.
  • Provided with a data sheet and handling instructions.
Catalog No. 50-004-51971