Learn More
Medchemexpress LLC Cloprostenol | 40665-92-7 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12131410MG
Encompass_Preferred
Cloprostenol is a synthetic prostaglandin F2α (PGF2α) analogue used as a luteolytic agent in research. It acts as a prostaglandin receptor agonist and is used to induce luteolysis and to study reproductive conditions such as open-cervix pyometra. CAS number 40665-92-7; molecular weight 424.92; molecular formula C22H29ClO6.
  • prostaglandin receptor agonist
  • induces luteolysis in animal models
  • suitable for reproductive and veterinary research
  • high purity suitable for analytical and in vitro studies
  • stable when stored per manufacturer recommendations
Catalog No. 50-002-32852