Learn More
Medchemexpress LLC Cloprostenol | 40665-92-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1213145MG
Encompass_Preferred
Cloprostenol is a prostaglandin F2α (PGF2α) analogue used in research as a luteolytic agent and for studies such as open-cervix pyometra.
  • acts as a prostaglandin F2α analogue and induces luteolysis.
  • white to off-white solid appearance suitable for analytical use.
  • characterized by HPLC, 1H NMR, and MS to confirm identity and purity.
  • long-term storage stable at room temperature for the solid form; solvent storage conditions vary.
Catalog No. 50-002-31033