Learn More
Cayman Chemical Cloprostenol isopropyl este
SDP
Supplier:  Cayman Chemical 1001001610 MG
Encompass_Preferred
A synthetic analog of PGF2α; an FP receptor agonist and a potent luteolytic agent in rats and hamsters; the optically active, 15(R) enantiomer of cloprostenol responsible for the majority of its biological activity; a more lipid soluble form of cloprostenol
Catalog No. 50-288-8571