Learn More
Cayman Chemical Cloprostenol methyl ester
SDP
Supplier:  Cayman Chemical 1001011510 MG
Encompass_Preferred
A more lipid soluble form of (+)-cloprostenol, amenable for certain formulations
Catalog No. 50-289-0525