Learn More
Medchemexpress LLC Clopyralid | 1702-17-6 | MFCD00078655 | 97.0% | 192.00 g/mol | C6H3Cl2NO2 | 10 G

Supplier: Medchemexpress LLC HYW01874910G
Clopyralid is a pyridine-based herbicide provided as an analytical standard for research and environmental analysis. It is 3,6-dichloropyridine-2-carboxylic acid (CAS 1702-17-6), appearing as a stable crystalline solid used as a reference material in chromatography, residue testing, and method development.
- Analytical standard for research and environmental testing.
- CAS number 1702-17-6.
- Chemical formula C6H3Cl2NO2.
- Molecular weight 192.00 g/mol.
- Purity typically reported near 97% (verify lot specification).
- Solid crystalline form, stable under normal laboratory storage.
- Intended for analytical and research use only, not for clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.