Learn More
Medchemexpress LLC Clopyralid | 1702-17-6 | MFCD00078655 | 98.5% | 192.00 | C6H3Cl2NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW018749100G
Encompass_Preferred
Clopyralid is a selective pyridine-carboxylic herbicide (CAS 1702-17-6) supplied as a research-grade chemical for study and formulation development. It is used to control broadleaf weeds, including thistles and clovers, and is noted for its persistence in soils; users should follow handling and safety guidance in the SDS.
  • Selective control of broadleaf weeds.
  • High purity suitable for research (98.5%).
  • Defined chemical identity: C6H3Cl2NO2, MW 192.00.
  • Available in multiple laboratory pack sizes.
  • Documentation available including SDS, COA, and datasheet.
Catalog No. 50-003-00032