Learn More
Medchemexpress LLC Clopyralid | 1702-17-6 | MFCD00078655 | 98.5% | 192.00 g/mol | C6H3Cl2NO2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01874950G
Encompass_Preferred
Clopyralid (CAS 1702-17-6) is a selective picolinic acid-class herbicide provided as a high-purity solid for research and analytical use. It serves as a reference material for studies of herbicidal activity, environmental persistence, and ionic-liquid formation to modify efficacy and soil leaching behavior.
  • High purity suitable for analytical applications.
  • Solid powder form for laboratory handling and formulation studies.
  • Validated chemical identity and molecular weight for reference use.
  • Relevant for studies of herbicidal efficacy and environmental fate.
  • Can form ionic liquids to investigate reduced soil leaching and altered activity.
  • Available in multiple pack sizes to support diverse research needs.
Catalog No. 50-003-01363