Learn More
Medchemexpress LLC Cloransulam-methyl | 147150-35-4 | 99.4% | 429.81 g/mol | C15H13ClFN5O5S | 1MG

Supplier: Medchemexpress LLC HY1261761MG
Cloransulam-methyl is a triazolopyrimidine sulfonanilide herbicide supplied as an analytical standard for research and analytical applications. It acts as an acetolactate synthase (ALS) inhibitor and is used in herbicidal activity studies and residue analysis. The material is provided at high purity and packaged in small quantities appropriate for standards and method development.
- High purity suitable for analytical use (99.44%).
- Molecular formula C15H13ClFN5O5S, molecular weight 429.81 g·mol⁻¹.
- Intended for research and analytical applications, including residue analysis.
- Available in milligram pack sizes for preparing stock solutions.
- Soluble in DMSO for stock solution preparation.
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.