Learn More
Medchemexpress LLC Cloransulam-methyl | 147150-35-4 | 99.4% | 429.81 g·mol⁻¹ | C15H13ClFN5O5S | 50MG

Supplier: Medchemexpress LLC HY12617650MG
Cloransulam-methyl is a triazolopyrimidine sulfonanilide herbicide and analytical research standard that inhibits acetolactate synthase (ALS). It is used for studying ALS inhibition and for analytical testing related to herbicide residues and mode-of-action research. The solid material has documented solubility and storage profiles for laboratory use.
- Inhibits acetolactate synthase (ALS), useful for biochemical studies.
- Supplied as an analytical research standard suitable for laboratory testing.
- High purity appropriate for analytical and experimental applications.
- Soluble in DMSO at 16.67 mg/mL with ultrasonic warming to 60°C.
- Powder storage stability: -20°C for up to 3 years; defined solution stability at low temperatures.
- Molecular weight 429.81 g·mol⁻¹; formula C15H13ClFN5O5S.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.