Learn More
Cayman Chemical CLOrcyclIzInhydroCLOrID 500mg
SDP
Supplier:  Cayman Chemical 19239500 MG
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 9 nM) that is effective against HCV (EC50 = 44 nM), preventing viral entry into host cells
Catalog No. 50-298-3619