Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | MFCD00052012 | 99.92% | C13H16Cl3NO | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14197100MG
Encompass_Preferred
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It is structurally related to Pargyline and has minimal effect on the amounts of conjugated dopamine (DA) in the superfusate of rat striatum slices. This compound contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Irreversible and selective inhibitor: targets monoamine oxidase A (MAO-A).
  • Intended for scientific research.
  • Alkyne group allows for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • High purity at 99.92%.
  • Offered in various quantities, including solid and pre-dissolved solutions.
Catalog No. 50-003-53294