Learn More
Apexbio Technology LLC Clorprenaline HCL 6933-90-0 50mg
SDP
Supplier:  Apexbio Technology LLC B171350
Encompass_Preferred
Clorprenaline hydrochloride (CAS 6933-90-0) is a selective 2-adrenergic receptor agonist By stimulating 2-adrenergic receptors in bronchial smooth muscle it promotes muscle relaxation and airway dilation This pharmacological profile supports its use in studies investigating bronchial physiology mechanisms of bronchial smooth muscle modulation and the effects of 2-adrenergic agonists in respiratory research Clorprenaline HCl is commonly utilized as a tool compound to explore 2-adrenergic signaling pathways and evaluate novel therapeutic strategies for airway constriction and related disorders
Catalog No. 50-000-94726