Learn More
Medchemexpress LLC Clorsulon | 60200-06-8 | 99.99% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0488100MG
Encompass_Preferred
Clorsulon is an orally active flukicidal agent. It works by inhibiting glycolysis, the primary energy production pathway in flukes. It also acts as a competitive inhibitor of 3-phosphoglycolate and ATP, thereby inhibiting glucose utilization and the formation of acetate and propionate by mature *Fasciola hepatica* in vitro. It is used in studies of liver fluke (*Fasciola hepatica* and *Fasciola gigantica*) infection in calves and sheep.
  • Inhibits glycolysis in flukes
  • Competitive inhibitor of 3-phosphoglycolate and ATP
  • Inhibits glucose utilization and formation of acetate and propionate by mature *Fasciola hepatica*
  • Used in studies of liver fluke infection
Catalog No. 50-208-2560