Learn More
Selleck Chemical LLC Closantel Sodium S4105-1g
SDP
Supplier:  Selleck Chemical LLC S41051G
Encompass_Preferred
Closantel is a gram positive antibacterial activity inhibitor inhibiting the KinA/Spo0F system with IC50 of 3 8 M
Catalog No. 50-310-8882