Learn More
ATCC Clostridioides difficile
Supplier:  ATCC 43255
Encompass
Clostridioides difficile strain VPI 10463 was isolated from an abdominal wound. This strain is toxigenic (toxinotype 0) and was confirmed by PCR to contain the tcdA and tcdB genes. The whole-genome sequenced bacterial strain has applications in enteric disease research and infectious disease research.
Catalog No. 50-238-4160