Learn More
ATCC Clostridioides difficile
Supplier:  ATCC 700057
Encompass
Clostridioides difficile strain VPI 11186 [6] is a whole-genome sequenced bacterial strain that can be used in susceptibility testing. This strain was confirmed by PCR to be negative for the cdtB, tcdA, and tcdB genes.
Catalog No. 50-238-4903