Learn More
ATCC Clostridioides difficile; 90556-M6S
Supplier:  ATCC 9689
Encompass
Clostridioides difficile strain 90556-M6S [L.S. McClung 1780] is a whole-genome sequenced bacterial type strain with applications in enteric and infectious disease research. This strain is confirmed to have the tcdA and tcdB genes.
Catalog No. 50-238-2892