Learn More
ATCC Clostridium acetobutylicum
Supplier:  ATCC 824
Encompass
Clostridium acetobutylicum is a whole-genome sequenced type strain. This bacterial culture degrades xylan and produces acetone, acidolysin, butyl alcohol [butanol], autobacteriocin, and restriction endonuclease Cac8241.
Catalog No. 50-238-2686