Learn More
ATCC Clostridium innocuum; Strain B-3
Supplier:  ATCC 14501
Encompass
Clostridium innocuum strain B-3 is a bacterial type strain that was isolated from an appendiceal abscess. This strain is propagated anaerobically.
Catalog No. 50-238-2877