Learn More
ATCC Clostridium perfringens; Strain S 107
Supplier:  ATCC 13124
Encompass
Clostridium perfringens strain NCTC 8237 [ATCC 19408, CIP 103 409, CN 1491, NCIB 6125, NCTC 6125, S 107] is a whole-genome sequenced bacterial type strain. This toxigenic culture has applications in agricultural research, bioinformatics, quality control, media testing, and water testing.
Catalog No. 50-238-3774