Learn More
ATCC Clostridium septicum; 6 x 0.3mL
Supplier:  ATCC 12464MINIPACK
Encompass
ATCC 12464-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 12464™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-5120
 Restricted Item