Learn More
Medchemexpress LLC Clotrimazole | 23593-75-1 | 99.9% | 344.84 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY1088210MM/1ML
Encompass_Preferred
Clotrimazole is an imidazole derivative, an antifungal compound and a CYP (cytochrome P450) inhibitor. It also possesses antibacterial activity.
  • Antifungal compound
  • CYP (cytochrome P450) inhibitor
  • Antibacterial activity
Catalog No. 50-003-44590