Learn More
Selleck Chemical LLC Clotrimazole S1606-1g
SDP
Supplier:  Selleck Chemical LLC S16061G
Encompass_Preferred
Clotrimazole (BAY b 5097 FB 5097) alters the permeability of the fungal cell wall by inhibiting the biosynthesis of ergosterol used in the treatment of fungal infections
Catalog No. 50-311-0905