Learn More
Medchemexpress LLC Clotrimazole-d10 | 00-00-0 | 97.5% | 354.90 g/mol | C22H7D10ClN2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10882S15MG
Encompass_Preferred
Clotrimazole-d10 is the deuterated isotopologue of clotrimazole supplied as a purified research reagent for analytical and tracer studies. It is intended for laboratory use only and not for human or clinical applications.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for analytical applications (97.5%).
  • Molecular weight 354.90 g/mol; formula C22H7D10ClN2.
  • Supplied in a 5 mg quantity for small-scale studies.
  • Accompanied by batch-specific certificate of analysis for traceability.
Catalog No. 50-002-64654