Learn More
Sigma Aldrich Fine Chemicals Biosciences Clotrimazole Related Compo50MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PHR270050MG
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Catalog No. 50-290-2141