Learn More
Cole-Parmer Digi-Sense ASTM Like Liquid-In-Glass Thermometer, 6C / Low Cloud and Pour, 76mm Immersion, -80 to 20C, Organic Liquid Fill
Supplier:  Cole-Parmer 0800905
Encompass_Preferred
These thermometers are precision manufactured to conform to ASTM E-1 specifications except for not being mercury filled. Designed for use in quality control laboratories or when mercury filled thermometers can not be used but where ASTM standards are still needed. ASTM established thermometer standards suited for special applications, and assigned an ASTM number according to each specification. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.
Catalog No. NC2168083
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov