Learn More
Medchemexpress LLC Cloxacillin sodium | 642-78-4 | 99.1% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0466B10MM/1ML
Encompass_Preferred
Cloxacillin sodium is an orally active antibacterial agent and a β-lactamase inhibitor with an IC50 of 0.04 μM. It can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB, and NLRP3-related proteins.
  • Orally active antibacterial agent
  • β-lactamase inhibitor
  • Inhibits S. aureus growth with MIC values of 0.125 μg/mL
  • Suppresses inflammatory response by inhibiting activation of MAPKs, NF-κB, and NLRP3-related proteins
  • Potential for treating bacterial infections, especially those caused by S. aureus
  • Offers a mechanism to combat bacterial resistance through β-lactamase inhibition
Catalog No. 50-004-07726