Learn More
Medchemexpress LLC Cloxiquine (5-Chloro-8-quinolinol) | 130-16-5 | 99.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB0963
Encompass_Preferred
Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal, and antiamoebic agent. It can be used for the research of tuberculosis and dermatoses, and suppresses the growth and metastasis of melanoma cells through the activation of PPARγ.
  • Antibacterial, antifungal, and antiamoebic properties
  • Suitable for tuberculosis and dermatoses research
  • Suppresses growth and metastasis of melanoma cells
  • Exhibits antituberculosis activities with low MICs
  • Inhibits migration of B16F10 and A375 cells
  • Suppresses glycolysis in B16F10 cells
  • Proven to suppress tumor growth in mouse models
  • Demonstrated to suppress tumor metastasis in mouse models
Catalog No. 50-004-06585