Learn More
Cayman Chemical ClozapIn-d4 100ug
SDP
Supplier:  Cayman Chemical 17513100 UG
Encompass_Preferred
An internal standard for the quantification of clozapine by GC- or LC-MS
Catalog No. 50-298-0214