Learn More
Medchemexpress LLC Clozapine-d8 | 1040850-25-1 | MFCD00223707 | >98% | C18H11D8ClN4 | 1 MG
Supplier: Medchemexpress LLC HY14539S1MG
Clozapine-d8 is a deuterium-labeled clozapine. It is an antipsychotic used for the research of schizophrenia, exhibiting high affinity for various neuroreceptors.
- Antipsychotic used for schizophrenia research.
- High affinity for several neuroreceptors.
- Potent antagonist of dopamine D2.
- Inhibits muscarinic M1 receptor and serotonin 5HT2A receptor.
- Potent and selective agonist at muscarinic M4 receptor.