Learn More
Enzo Life Sciences Clozapine N-oxide, (25mg), CAS Number: 34233-69-7
SDP
Supplier:  Enzo Life Sciences BMLNS1050025
Encompass_Preferred
Major metabolite of clozapine. Major metabolite of clozapine. Possesses little or no activity at 5-HT receptors. Formula: C18H19ClN4O . CH3OH. MW: 342.8 . 32.0. CAS Number: 34233-69-7. Purity: ≥99% (HPLC). Appearance: Yellow solid. Solubility: Soluble in water (5mg/ml). Use/Stability: Store solutions at -20°C for up to 4 months. Long Term Storage: Ambient
Catalog No. 50-103-1781