Learn More
Medchemexpress LLC Clozapine (Standard) | 5786-21-0 | 99.84% | C18H19ClN4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14539R
Encompass_Preferred
Clozapine (Standard) is an analytical standard intended for research and analytical applications. As an antipsychotic, it is used for the research of schizophrenia and exhibits high affinity for several neuroreceptors. Clozapine acts as a potent antagonist of dopamine D2 (Ki of 75 nM), and inhibits muscarinic M1 and serotonin 5HT2A receptors (Kis of 9.5 nM and 4 nM, respectively). It also functions as a potent and selective agonist at the muscarinic M4 receptor (EC50=11 nM).
  • Potent antagonist of dopamine D2
  • Inhibits muscarinic M1 and serotonin 5HT2A receptors
  • Potent and selective agonist at the muscarinic M4 receptor
  • Used in schizophrenia research
  • Intended for research and analytical applications
Catalog No. 50-003-54745